Learn More
DDX4 VASA Mouse anti-Human, Unconjugated, Clone: 1F12C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67147-2-IG-150UL
 View more versions of this product
Product Code. 16839615
  • 4652.90 SEK / 150µL
Description
The DDX4, VASA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX4, VASA. This antibody recognizes human, mouse antigen. The DDX4, VASA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
DDX4 VASA
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry
1F12C3
Unconjugated
150 ÎĽL
54514
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DDX4 VASA Mouse anti-Human, Unconjugated, Clone: 1F12C3, Proteintech > 150 ÎĽL