Learn More
DDX4 VASA Mouse anti-Human, Unconjugated, Clone: 1F12C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67147-2-IG-150UL
Description
The DDX4, VASA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDX4, VASA. This antibody recognizes human, mouse antigen. The DDX4, VASA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
DDX4 VASA | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Immunohistochemistry | |
1F12C3 | |
Unconjugated | |
150 ÎĽL | |
54514 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.