missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11369-1-AP-20UL
This item is not returnable.
View return policy
Description
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.Specifications
DEF6 | |
Polyclonal | |
Unconjugated | |
DEF6 | |
DEF 6, DEF6, IBP, IRF4 binding protein, SLAT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23853, 50619 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C2K1, Q9H4E7 | |
DEF6 | |
DEF6 Fusion Protein Ag1925 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DEF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction