missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dermatopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14777-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.Specifications
Dermatopontin | |
Polyclonal | |
Unconjugated | |
DPT | |
Dermatopontin, DPT, TRAMP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1805, 289178, 56429 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q07507, Q9QZZ6 | |
Dpt | |
Dermatopontin Fusion Protein Ag6200 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |