missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11561-1-AP-150UL
This item is not returnable.
View return policy
Description
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.Specifications
DGAT1 | |
Polyclonal | |
Unconjugated | |
DGAT1 | |
ACAT related gene product 1, AGRP1, ARGP1, DGAT, DGAT1, Diglyceride acyltransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13350, 84497, 8694 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75907, Q9ERM3, Q9Z2A7 | |
Dgat1 | |
DGAT1 Fusion Protein Ag2123 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction