Learn More
DHCR24 Mouse anti-Human, Unconjugated, Clone: 2F6E11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60028-1-IG-150UL
 View more versions of this product
Product Code. 16886975
  • 4700.76 SEK / 150µL
Description
The DHCR24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DHCR24. This antibody recognizes human antigen. The DHCR24 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
DHCR24
Monoclonal
1080 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
2F6E11
Unconjugated
150 μL
1718
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DHCR24 Mouse anti-Human, Unconjugated, Clone: 2F6E11, Proteintech > 150 μL