Learn More
DHCR24 Mouse anti-Human, Unconjugated, Clone: 2F6E11, Proteintech
Mouse Monoclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | DHCR24 |
---|---|
Clone | 2F6E11 |
Concentration | 1080 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896975
|
Proteintech
60028-1-IG-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886975
|
Proteintech
60028-1-IG-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DHCR24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DHCR24. This antibody recognizes human antigen. The DHCR24 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
DHCR24 | |
1080 ÎĽg/mL | |
Monoclonal | |
Mouse | |
1718 | |
Primary |
2F6E11 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.