Learn More
DHCR24 Mouse anti-Human, Unconjugated, Clone: 2F6E11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60028-1-IG-150UL
 View more versions of this product
Product Code. 16886975
  • 4700.76 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The DHCR24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DHCR24. This antibody recognizes human antigen. The DHCR24 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
DHCR24
Monoclonal
1080 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
2F6E11
Unconjugated
150 μL
1718
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DHCR24 Mouse anti-Human, Unconjugated, Clone: 2F6E11, Proteintech > 150 μL