missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIDO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10183-1-AP-20UL
This item is not returnable.
View return policy
Description
The immunogen of this antibody is the C-terminal of isoform 2 of DATF1 (DIDO1).
Dio-1 (Death Inducer-Obliterator-1) is a putative transcription factor that contains two zinc finger motifs. Dio-1 translocates to the nucleus, and activates apoptosis during limb development. Programmed cell death, a highly regulated form of apoptosis, plays an important role in determining the amount of tissue, the shape, and the definition of each digit during limb development. Dio-1 expression is upregulated when an apoptotic signal is detected, and subsequently apoptosis is induced. This process is similar to the expression of NFkB and NGF in response to external signals. Dio-1 expression is suppressed by caspase inhibitors and Bcl-2 expression. This supports the theory that Dio-1 functions in the onset of programmed cell death.Specifications
DIDO1 | |
Polyclonal | |
Unconjugated | |
DIDO1 | |
BYE1, C20orf158, DATF 1, DATF1, death inducer obliterator 1, DIDO1, DIDO2, DIDO3, DIO 1, DIO1, hDido1, KIAA0333 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11083, 23856, 362286 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Flow Cytometry | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C9B9, Q9BTC0 | |
Dido1 | |
DIDO1 Fusion Protein Ag0237 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DIDO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction