missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIDO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DIDO1 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886393
|
Proteintech
10183-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876393
|
Proteintech
10183-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The immunogen of this antibody is the C-terminal of isoform 2 of DATF1 (DIDO1).
Dio-1 (Death Inducer-Obliterator-1) is a putative transcription factor that contains two zinc finger motifs. Dio-1 translocates to the nucleus, and activates apoptosis during limb development. Programmed cell death, a highly regulated form of apoptosis, plays an important role in determining the amount of tissue, the shape, and the definition of each digit during limb development. Dio-1 expression is upregulated when an apoptotic signal is detected, and subsequently apoptosis is induced. This process is similar to the expression of NFkB and NGF in response to external signals. Dio-1 expression is suppressed by caspase inhibitors and Bcl-2 expression. This supports the theory that Dio-1 functions in the onset of programmed cell death.Specifications
DIDO1 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q8C9B9, Q9BTC0 | |
11083, 23856, 362286 | |
DIDO1 Fusion Protein Ag0237 | |
Primary | |
-20°C | |
DIDO1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BYE1, C20orf158, DATF 1, DATF1, death inducer obliterator 1, DIDO1, DIDO2, DIDO3, DIO 1, DIO1, hDido1, KIAA0333 | |
Dido1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DIDO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech