Learn More
Scientific Industries SI™ Disruptor Genie™ Analog Cell Disruptor
Increases the disruption efficiency of disruption beads with unique, patented microtube holder design. Scientific Industries SI™ Disruptor Genie™ Analog Cell Disruptor provides vigorous mechanical cell disruption.
Brand:  Scientific Industries SI™ SI-D278
7612.38 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15567345
  • 8000.00 SEK / Each
Only null left
Estimated Shipment: 14-08-2025
Description
  • Rapid hands-free disruption of up to 12 microtubes (1.5mL or 2.0mL)
  • Supplied with NEW Universal Microtube Holder
  • Set timer to run for up to 15 minutes or continuously
  • Suitable for use in cold rooms or incubators
  • Less costly than ultrasonic units or homogenizers
Applications include:
  • Disruption of bacterial cultures for nucleic acid isolation and downstream processing
  • Grinding plant tissue for DNA extractions
  • Preparation of cell lysates from yeast or fungi
  • Rapid resuspension of pellets
  • Dissolution of chemical substances
Specifications
Specifications
2850 rpm
230 V
1.5/2 mL
50 Hz
190 mm
4.3 kg
Universal microtube holder
230 V, 50 Hz
Microtubes
CH Plug
165 mm
122 mm
SI-D278
12
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Scientific Industries SI™ Disruptor Genie™ Analog Cell Disruptor >