missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21112-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.Specifications
DKK1 | |
Polyclonal | |
Unconjugated | |
DKK1 | |
Dickkopf 1, Dickkopf related protein 1, DKK 1, DKK1, hDkk 1, SK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13380, 22943, 293897 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54908, O94907 | |
Dkk1 | |
DKK1 Fusion Protein Ag15110 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DKK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction