missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18080-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to DNAH6.
DNAH6 is a protein coding gene.Specifications
DNAH6 | |
Polyclonal | |
Unconjugated | |
DNAH6 | |
Ciliary dynein heavy chain 6, DNAH6, Dnahc6, DNHL1, Dynein heavy chain 6, axonemal, HL 2, HL2, KIAA1697 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117250, 1768, 330355 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9C0G6 | |
Dnah6 | |
DNAH6 Fusion Protein Ag12774 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNAH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction