missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15934-1-AP-150UL
This item is not returnable.
View return policy
Description
DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codomit alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.Specifications
DNASE2 | |
Polyclonal | |
Unconjugated | |
DNASE2 | |
Acid DNase, Deoxyribonuclease 2 alpha, Deoxyribonuclease II alpha, DNase II alpha, DNASE2, DNASE2A, DNL, DNL2, Lysosomal DNase II, R31240_2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13423, 171575, 1777 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00115, P56542, Q9QZK8 | |
Dnase2 | |
DNASE2 Fusion Protein Ag8781 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNASE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction