Learn More
DOCK7 Mouse, anti-Human, Unconjugated, Clone: 1A4A6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67842-1-IG-20UL
Description
DOCK 7 (dedicator of cytokinesis 7), also known as ZIR2, is a member of the DOCK180-related protein superfamily. Expressed mainly in neuronal cells, DOCK 7 is a guanine nucleotide exchange factor (GEF) for small GTPases, Rac1 and Cdc42, which are the major regulators of actin cytoskeleton. Multiple isoforms of DOCK 7 exist due to alternative splicing events.
Specifications
DOCK7 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat |
Western Blot, ELISA | |
1A4A6 | |
Unconjugated | |
20 μL | |
85440 | |