missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20310-1-AP-20UL
This item is not returnable.
View return policy
Description
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.Specifications
DRD5 | |
Polyclonal | |
Unconjugated | |
DRD5 | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, DBDR, Dopamine D5 receptor, dopamine receptor D5, DRD1B, DRD1L2, DRD5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13492, 1816, 25195 | |
-20°C | |
Liquid |
Western Blot, Flow Cytometry | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21918, P25115, Q8BLD9 | |
Drd5 | |
DRD5 Fusion Protein Ag14145 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DRD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction