missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drebrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25770-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer′s disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.Specifications
Drebrin | |
Polyclonal | |
Unconjugated | |
DBN1 | |
D0S117E, DBN1, Drebrin, drebrin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1627, 56320 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16643, Q9QXS6 | |
Dbn1 | |
Drebrin Fusion Protein Ag22634 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Drebrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction