Learn More
DSE Mouse anti-Human, Mouse, Clone: 2C9F7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60041-1-IG-150UL
 View more versions of this product
Product Code. 16886985
  • 4700.76 SEK / 150µL
Description
Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
Specifications
Specifications
DSE
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BLI4, Q9UL01
Dse
DSE Fusion Protein Ag0695
150 μL
Primary
Human, Mouse
Antibody
IgM
Immunohistochemistry (Paraffin), Western Blot
2C9F7
Unconjugated
DSE
dermatan sulfate epimerase, DS epimerase, DSE, DSEPI, SART 2, SART2
Mouse
Caprylic Acid Precipitation
RUO
212898, 29940
-20°C
Liquid
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DSE Mouse anti-Human, Mouse, Clone: 2C9F7, Proteintech > 150 μL; Unconjugated