missing translation for 'onlineSavingsMsg'
Learn More

DSE Mouse anti-Human, Mouse, Clone: 2C9F7, Proteintech

Mouse Monoclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen DSE
Clone 2C9F7
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896985
View Documents
Proteintech
60041-1-IG-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16886985
View Documents
Proteintech
60041-1-IG-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
Specifications

Specifications

DSE
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
dermatan sulfate epimerase, DS epimerase, DSE, DSEPI, SART 2, SART2
Dse
IgM
Caprylic Acid Precipitation
Antibody
2C9F7
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Mouse, Human
Q8BLI4, Q9UL01
212898, 29940
DSE Fusion Protein Ag0695
Primary
-20°C
DSE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DSE Mouse anti-Human, Mouse, Clone: 2C9F7, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.