Learn More
DWK Life Sciences DURAN™ Multi-purpose Cylinder, with round base, without graduation
Cylinders without graduation can be used to mix components.
458.00 SEK - 8305.00 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Diluting solutions; Mixing several components |
Autoclavable | Yes |
Quantity | 10/Pk. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11746998
|
DWK Life Sciences
213986806 |
380 mL |
2820.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15201729
|
DWK Life Sciences
213983406 |
180 mL |
1875.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15804508
|
DWK Life Sciences
213982101 |
220 mL |
2290.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15231729
|
DWK Life Sciences
213985201 |
280 mL |
2250.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15211729
|
DWK Life Sciences
213983603 |
420 mL |
2870.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15221729
|
DWK Life Sciences
213984608 |
530 mL |
3180.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15241729
|
DWK Life Sciences
213985304 |
450 mL |
2715.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15251729
|
DWK Life Sciences
213987408 |
1650 mL |
8305.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10334261
|
DWK Life Sciences
213988001 |
770 mL |
458.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15261729
|
DWK Life Sciences
213987708 |
1250 mL |
4270.00 SEK
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Rough ground rim
- With round base
Specifications
Borosilicate Glass (3.3) | |
Diluting solutions; Mixing several components | |
Yes | |
10/Pk. |