Learn More
DWK Life Sciences DURAN™ Three-way Stopcock, complete with PTFE key, T-bore
3-way tap with PTFE plug for grease free operation.
366.52 SEK - 2915.00 SEK
Specifications
Material | Borosilicate Glass (3.3), PTFE |
---|---|
Autoclavable | Autoclavable |
Width (Metric) | 1.5 mm |
No. per Pack | 5/Pk. |
For Use With (Application) | Installation in glass apparatus and devices |
Beschreibung
T-valves are the most common form of 3-way valves. The DURAN™ three-way valve offers the following adjustment options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments The polished version with PTFE plug allows grease free working.
- 3-way valve with PTFE valve plug
- PTFE threaded tap plug with T-port
- Grease-free working
- Three-way valve offers the following setting options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
- Glass articles for further processing
Spezifikation
Borosilicate Glass (3.3), PTFE | |
1.5 mm | |
Installation in glass apparatus and devices |
Autoclavable | |
5/Pk. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.