missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamitin (69-376 AA) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10030-2-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is against the internal region (69-376aa) of full length p50 dynamitin.
This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kDa. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place.Specifications
Dynamitin (69-376 AA) | |
Polyclonal | |
Unconjugated | |
DCTN2 | |
DCTN 50, DCTN2, DCTN50, dynactin 2, dynactin 2 (p50), Dynactin subunit 2, DYNAMITIN, p50 dynamitin, RBP50 | |
Rabbit | |
20 ÎĽL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13561, Q99KJ8 | |
Dctn2 | |
Antigen Affinity Chromatography | |
RUO | |
10540, 69654 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dynamitin (69-376 AA) Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction