missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2AK2 Mouse anti-Human, Clone: 2A8A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66646-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to EIF2AK2.
PKR is a 68 kDa protein and an IFN-induced serine/threonine protein kinase consisting of two N-terminal RNA-binding regulatory domains and a functional C-terminal serine/threonine protein kinase domain. Activated by dsRNA produced during viral infections, PKR plays a key role in IFN induced-innate antiviral response, virus-induced apoptosis, cell growth & differentiation. Upon activation, PKR gets autophosphorylated and catalyzes the phosphorylation of elF2 alpha subunit, which leads to an inhibition of protein synthesis. PKR induces apoptosis by up-regulating Fas expression and mediates FADD/Caspase-8 death signaling pathway. Upon viral infection, it functions as a dual protein, sequentially activating both cell survival & cell death pathways using kinase independent and dependent strategies. PRKR regulates multiple pathways which include NF-kappaB activation, p53, p38, & PDGF signaling pathway. PKR has been implicated in tumor suppression & maligcy. To evade the antiviral effects of PKR, viruses have evolved multiple mechanisms, such as the inhibition of PKR by the non-structural protein (NS1) of the influenza virus. More recently, PKR has been implicated in several neurodegenerative diseases including Alzheimer, Huntington, and amyotrophic lateral sclerosis.Specifications
EIF2AK2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19525 | |
EIF2AK2 | |
EIF2AK2 Fusion Protein Ag13010 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
2A8A4 | |
Unconjugated | |
EIF2AK2 | |
eIF 2A protein kinase 2, EIF2AK1, EIF2AK2, P1/eIF 2A protein kinase, p68 kinase, PKR, PRKR, Protein kinase RNA activated | |
Mouse | |
Protein G | |
RUO | |
5610 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EIF2AK2 Mouse anti-Human, Clone: 2A8A4, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction