Learn More
EIF5A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11309-1-AP-20UL
Description
The EIF5A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EIF5A. This antibody recognizes human, mouse, rat antigen. The EIF5A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
EIF5A | |
Polyclonal | |
Unconjugated | |
20 μL | |
1984 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
420 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.