Learn More
EIF5A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | EIF5A |
---|---|
Concentration | 420 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888293
|
Proteintech
11309-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878293
|
Proteintech
11309-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EIF5A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EIF5A. This antibody recognizes human, mouse, rat antigen. The EIF5A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
EIF5A | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
420 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
1984 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.