Learn More
EIF5A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11309-1-AP-150UL
 View more versions of this product
Product Code. 16878293
  • 4652.90 SEK / 150µL
Description
The EIF5A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EIF5A. This antibody recognizes human, mouse, rat antigen. The EIF5A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
EIF5A
Polyclonal
Unconjugated
150 μL
1984
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
420 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
EIF5A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL