Learn More
Brady™ Electrostatic Dissipative (ESD) Polyester Circuit Board Labels for M6/M7 Printers
WorkHorse™ Glossy White Static Dissipative Circuit Board Labels is ideal for barcode, printed circuit board and electronic Equipment Identification.
Brand:  Brady™ 174192
 View more versions of this product
Product Code. 17848244
  • 888.00 SEK / Pack of 750
Only null left
Estimated Shipment: 20-08-2025
Description
  • Meets recommended range for Dissipative ESD packaging materials as defined by ANSI/ESD S541-2008
  • Meets UL969 Labeling and Marking Standard; CSA Accepted, RoHS Compliant
  • Has surface resistivity values in the recommended range for dissipative ESD packaging materials
  • Made of B-473 polyester material
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ Electrostatic Dissipative (ESD) Polyester Circuit Board Labels for M6/M7 Printers > Polyester Circuit Board Labels; Dimensions (LxW): 0.25 x 0.25 in.; For Use With (Equipment): M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter); Height (Metric): 6.35 mm