Learn More
Brady™ Electrostatic Dissipative (ESD) Polyester Circuit Board Labels for M6/M7 Printers
WorkHorse™ Glossy White Static Dissipative Circuit Board Labels is ideal for barcode, printed circuit board and electronic Equipment Identification.
Brand:  Brady™ 174192
 View more versions of this product
Product Code. 17848244
  • 888.00 SEK / Pack of 750
Only null left
Estimated Shipment: 01-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Meets recommended range for Dissipative ESD packaging materials as defined by ANSI/ESD S541-2008
  • Meets UL969 Labeling and Marking Standard; CSA Accepted, RoHS Compliant
  • Has surface resistivity values in the recommended range for dissipative ESD packaging materials
  • Made of B-473 polyester material
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Electrostatic Dissipative (ESD) Polyester Circuit Board Labels for M6/M7 Printers > Polyester Circuit Board Labels; Dimensions (LxW): 0.25 x 0.25 in.; For Use With (Equipment): M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter); Height (Metric): 6.35 mm