missing translation for 'onlineSavingsMsg'
Learn More

ELF1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55029-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886465

  • 3665.00 SEK / 150µL
Estimated Shipment: 30-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to ELF1.

This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

ELF1
Polyclonal
Unconjugated
ELF1
E74 like factor 1, ELF1
Rabbit
Antigen Affinity Chromatography
RUO
1997
-20°C
Liquid
Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P32519
ELF1
Peptide
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers