missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12499-1-AP-20UL
This item is not returnable.
View return policy
Description
The ELF2 gene encodes a protein that physically interacts with AML1 and mediates opposing effects on AML1-mediated transcription of the B cell-specific blk gene.Specifications
ELF2 | |
Polyclonal | |
Unconjugated | |
ELF2 | |
E74 like factor 2, ELF2, ELF2A, EU32, NERF, NERF 1A, NERF 1a,b, NERF 1B, NERF 2, New ETS related factor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1998, 361944, 69257 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15723, Q9JHC9 | |
Elf2 | |
ELF2 Fusion Protein Ag3170 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ELF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction