missing translation for 'onlineSavingsMsg'
Learn More

ELF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen ELF2
Concentration 0.2 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16890244
View Documents
Proteintech
12499-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16880244
View Documents
Proteintech
12499-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ELF2 gene encodes a protein that physically interacts with AML1 and mediates opposing effects on AML1-mediated transcription of the B cell-specific blk gene.
Specifications

Specifications

ELF2
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Rat, Human
Q15723, Q9JHC9
1998, 361944, 69257
ELF2 Fusion Protein Ag3170
Primary
-20°C
ELF2
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E74 like factor 2, ELF2, ELF2A, EU32, NERF, NERF 1A, NERF 1a,b, NERF 1B, NERF 2, New ETS related factor
Elf2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ELF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.