Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Brand: Eppendorf™ 5804787005
2327.27 SEK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Adapter | |
1 x 250 mL Tubes | |
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges | |
250 mL |
Product Content Correction
Product Title
Eppendorf™ S-4-72 Rotor Adapters > For 1 x 250mL tubes
Spot an opportunity for improvement?Share a Content Correction