Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Brand:  Eppendorf™ 5804785002
2451.27 SEK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 13286589
  • 2580.28 SEK / Pack of 2
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Description
  • For use with Centrifuge 5804/5804 R and 5810/5810 R
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
2 x 50 mL Conical Tubes
50 mL
Conical
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ S-4-72 Rotor Adapters > For 2 x 50mL conical tubes