missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERGIC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11927-1-AP-20UL
This item is not returnable.
View return policy
Description
Possible role in transport between endoplasmic reticulum and Golgi.Specifications
ERGIC2 | |
Polyclonal | |
Unconjugated | |
ERGIC2 | |
cd002, CDA14, ERGIC and Golgi 2, ERGIC2, Erv41, PTX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51290 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96RQ1 | |
ERGIC2 | |
ERGIC2 Fusion Protein Ag2603 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ERGIC2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction