missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESCO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23525-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that may have acetyltransferase activity and may be required for the establishment of sister chromatid cohesion during the S phase of mitosis. Mutations in this gene have been associated with Roberts syndrome.Specifications
ESCO2 | |
Polyclonal | |
Unconjugated | |
ESCO2 | |
2410004I17Rik, ECO1 homolog 2, EFO2, ESCO2, N acetyltransferase ESCO2, RBS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
157570, 71988 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q56NI9, Q8CIB9 | |
Esco2 | |
ESCO2 Fusion Protein Ag18893 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ESCO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction