missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESCO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ESCO2 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882375
|
Proteintech
23525-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872375
|
Proteintech
23525-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein that may have acetyltransferase activity and may be required for the establishment of sister chromatid cohesion during the S phase of mitosis. Mutations in this gene have been associated with Roberts syndrome.Specifications
ESCO2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q56NI9, Q8CIB9 | |
157570, 71988 | |
ESCO2 Fusion Protein Ag18893 | |
Primary | |
-20°C | |
ESCO2 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
2410004I17Rik, ECO1 homolog 2, EFO2, ESCO2, N acetyltransferase ESCO2, RBS | |
Esco2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ESCO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech