Learn More
ESD Mouse anti-Human, Unconjugated, Clone: 2F9A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67069-1-IG-20UL
 View more versions of this product
Product Code. 16889475
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
The ESD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ESD. This antibody recognizes human antigen. The ESD antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ESD
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2F9A7
Unconjugated
20 μL
2098
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ESD Mouse anti-Human, Unconjugated, Clone: 2F9A7, Proteintech > 20 μL