missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-bromopyruvate, tech. 75% Product Code.: 11340124

Ethyl 3-bromopyruvate, tech. 75%

Product Code. 11340124
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11340124

Brand: Thermo Scientific Alfa Aesar L00582.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-23-5 | C5H7BrO3 | 195.01 g/mol

Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide.

Solubility
Difficult to mix.

Notes
Moisture Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-23-5
Molecular Formula C5H7BrO3
Molecular Weight (g/mol) 195.01
MDL Number MFCD00000204
InChI Key VICYTAYPKBLQFB-UHFFFAOYSA-N
Synonym ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester
PubChem CID 66144
IUPAC Name ethyl 3-bromo-2-oxopropanoate
SMILES CCOC(=O)C(=O)CBr

Specifications

Density 1.555
Boiling Point 98°C to 100°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.469
Quantity 25 g
UN Number UN3265
Beilstein 1760158
Sensitivity Moisture sensitive
Solubility Information Difficult to mix.
Formula Weight 195.02
Percent Purity 75%
Grade Technical
Chemical Name or Material Ethyl 3-bromopyruvate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10241438 Certificate TypeCertificate of Analysis Date12/04/2024 Product CodeL00582.06,L00582.22,L00582.14
Lot Number10247227 Certificate TypeCertificate of Analysis Date05/07/2023 Product CodeL00582.06,L00582,L00582.22,L00582.14
Lot Number10222281 Certificate TypeCertificate of Analysis Date10/08/2022 Product CodeL00582.06,L00582.22,L00582.14
Lot Number10217956 Certificate TypeCertificate of Analysis Date21/05/2022 Product CodeL00582.06,L00582.22,L00582.14
Lot Number10182264 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL00582.06,L00582.22,L00582.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-bromopyruvate, tech. 75% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.