missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Monopropyl Ether, ReagentPlus™ 99.4%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass bottle
Unit Size:
1 L
4 L
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Density | 0.913g/mL at 25°C |
Packaging | Glass bottle |
Linear Formula | CH3CH2CH2OCH2CH2OH |
Refractive Index | 1.427 |
Quantity | 4 L |
UN Number | UN1993C |
Beilstein | 1731983 |
Formula Weight | 104.15g/mol |
Vapor Pressure | 1.3mmHg (20°C) |
Percent Purity | 99.45% |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylene Glycol Monopropyl Ether, ReagentPlus™ 99.4%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction