Learn More
Eutech pH Buffer Sachets
Get a wide range of Eutech™ pH buffers and solutions for use with pH meters and electrodes
1026.48 SEK - 1032.18 SEK
Product Code | Brand | Chemical Name or Material | pH | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Chemical Name or Material | pH | Price | Quantity & Availability | ||||
11563785
|
Eutech
01X223102 |
pH Buffer Sachet | 4.01 |
1032.18 SEK
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11553785
|
Eutech
01X223101 |
pH Buffer Sachet | 7 |
1032.18 SEK
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15691707
|
Eutech
ECBU4710R |
pH Buffer Sachet Set, Deionized Water | 4.01, 7.00, 10.01 |
1026.48 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
pH Buffer Sachets are NIST-traceable sachets for quick, accurate and convenient calibrations. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
- pH Buffer Sachets -20 ml
- NIST Traceable
Specifications
NIST Traceable |