Learn More
Lenz Laborglasintrumente Extractor Thimbles for Solids
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1190.00 SEK - 1235.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10070521
|
Lenz Laborglasintrumente
05431131 |
70 mL |
1190.00 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
10559581
|
Lenz Laborglasintrumente
05431137 |
100 mL |
1235.00 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
Description
- For solids
- For glass inserts for solids
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) | |
NS 45/40 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Thimbles for Solids