Learn More
FABP6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13781-1-AP-150UL
 View more versions of this product
Product Code. 16882164
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The FABP6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FABP6. This antibody recognizes human, mouse antigen. The FABP6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FABP6
Polyclonal
Unconjugated
150 μL
2172
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
333 μg/mL
Rabbit
Primary
Human, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FABP6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL