Learn More
FABP6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13781-1-AP-150UL
Description
The FABP6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FABP6. This antibody recognizes human, mouse antigen. The FABP6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
FABP6 | |
Polyclonal | |
Unconjugated | |
150 μL | |
2172 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |