missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM20C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25395-1-AP-20UL
This item is not returnable.
View return policy
Description
Calcium-binding protein which may play a role in dentin mineralization.Specifications
FAM20C | |
Polyclonal | |
Unconjugated | |
FAM20C | |
Dentin matrix protein 4, DMP 4, DMP4, FAM20C, Protein FAM20C, RNS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304334, 56975, 80752 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5MJS3, Q8IXL6 | |
FAM20C | |
FAM20C Fusion Protein Ag22061 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FAM20C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction