missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11161-1-AP-20UL
This item is not returnable.
View return policy
Description
FBXO9 (F-box protein 9) is a Ubl conjugation pathway protein and it contains one F-box domain and one TPR repeat. FBXO9 may act as a potential sepsis biomarker or therapeutic target. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which functions in phosphorylation-dependent ubiquitination.Specifications
FBXO9 | |
Polyclonal | |
Unconjugated | |
FBXO9 | |
F box only protein 9, F box protein 9, FBX9, FBXO9, KIAA0936, NY REN 57, VCIA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26268, 300849, 71538 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5U2X1, Q8BK06, Q9UK97 | |
Fbxo9 | |
FBXO9 Fusion Protein Ag1643 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FBXO9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction