missing translation for 'onlineSavingsMsg'
Learn More

FBXO9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen FBXO9
Concentration 0.2 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888043
View Documents
Proteintech
11161-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16878043
View Documents
Proteintech
11161-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FBXO9 (F-box protein 9) is a Ubl conjugation pathway protein and it contains one F-box domain and one TPR repeat. FBXO9 may act as a potential sepsis biomarker or therapeutic target. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which functions in phosphorylation-dependent ubiquitination.
Specifications

Specifications

FBXO9
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
Q5U2X1, Q8BK06, Q9UK97
26268, 300849, 71538
FBXO9 Fusion Protein Ag1643
Primary
-20°C
FBXO9
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
F box only protein 9, F box protein 9, FBX9, FBXO9, KIAA0936, NY REN 57, VCIA1
Fbxo9
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
FBXO9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.