missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fetuin, From Fetal Bovine Serum
Fetuin is a protein containing 12 Cys residues, all involved in disulfide bridges.
2586.40 SEK - 9275.00 SEK
Specifications
For Use With (Application) | Type 2 Diabetes Mellitus (T2DM) Studies, Cell Attachment and Proliferation |
---|---|
Formulation | Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7% |
Name | Fetuin Protein |
Purification Method | Ammonium Sulfate Fractionation |
Source | Fetal Bovine Serum |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11431752
|
MP Biomedicals™
02104874.1 |
100mg |
2586.40 SEK
100mg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11441752
|
MP Biomedicals™
02104874.2 |
250mg |
4494.40 SEK
250mg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11451752
|
MP Biomedicals™
0210487401 |
1g |
9275.00 SEK
1g |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Formulation: Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%
Applications:
Fetuin bovine is a Protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients.
Specifications
Type 2 Diabetes Mellitus (T2DM) Studies, Cell Attachment and Proliferation | |
Fetuin Protein | |
Fetal Bovine Serum |
Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7% | |
Ammonium Sulfate Fractionation | |
Powder |