Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Brand: Thermo Scientific™ 820049
48.76 SEK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
- Use with fiberboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Each | |
49 (7 x 7 Grid) |
Promotions
-
Multiple Manufacturers
Sample Preparation, Filtration and Storage
Get up to 80% off some of our most popular products.Promo Code: 24124Expires: 2025-08-31 Redemption Only
New Offer Save Now -
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24
New Offer Save Up to 92%