Learn More
ROBU VitraPOR™ Filter Crucible, 30 mL
Crucibles are marked with an individual number and allow a distinct sample.
Brand:  ROBU VitraPOR™ 2030C
 View more versions of this product
Product Code. 16883006
  • 1740.00 SEK / Pack of 10
Only null left
Estimated Shipment: 07-08-2025
Description
  • For analytical filtration in laboratory
  • Suitable glass adapters and rubber sleeves are available
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
30 mL
Coarse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ROBU VitraPOR™ Filter Crucible, 30 mL > Pore Size: Coarse