Learn More
Macherey-Nagel™ Pleated Filter MN 615 1/4
Use for chemical analysis in waste water treatment. Pleated Filter MN 615 1/4 is perfect for determination of dry matter.
Brand: Macherey-Nagel™ 531024
This item is not returnable.
View return policy
Description
- Chemical analysis in a waste water treatment plant
- Ideally suited to be used for analytical procedures
- Reproducible results
- Norm conforming filtration
- Quality "Made in Germany"
- Determination of dry matter - filtration for all N-parameters
Specifications
Pleated | |
MN 615 1/4 | |
240 mm | |
240 mm |
Product Content Correction
Product Title
Macherey-Nagel™ Pleated Filter MN 615 1/4 > Diameter: 240 mm
Spot an opportunity for improvement?Share a Content Correction