Learn More
Macherey-Nagel™ Pleated Filter MN 615 1/4
Use for chemical analysis in waste water treatment. Pleated Filter MN 615 1/4 is perfect for determination of dry matter.
361.00 SEK - 841.00 SEK
Specifications
Format Pleated
Grade MN 615 1/4
View More Specs
Products 4
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10392711
View Documents
Macherey-Nagel™
531015
150 mm
361.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
10792712
View Documents
Macherey-Nagel™
531024
240 mm
595.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
10342811
View Documents
Macherey-Nagel™
531018
185 mm
400.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
10423321
View Documents
Macherey-Nagel™
531032
320 mm
841.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
Description
Description
  • Chemical analysis in a waste water treatment plant
  • Ideally suited to be used for analytical procedures 
  • Reproducible results
  • Norm conforming filtration
  • Quality “Made in Germany”
  • Determination of dry matter - filtration for all N-parameters
Specifications
Specifications
Pleated
MN 615 1/4
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Pleated Filter MN 615 1/4