Learn More
3M™ First Base™ + Bump Cap
First Base™ + Bump Cap designed to provide limited head protection from bumps and minor lacerations from stationary objects. Patented flexible shell design manufactured from durable ABS plastic provides industrial bump protection to EN812.
Brand:  3M™ 7100206561
2906.52 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17287680
  • 5075.00 SEK / Pack of 20
Only null left
Estimated Shipment: 16-09-2025
Description
  • Provides industrial bump protection to EN812.
  • Flexible shell made with ABS plastic, can be removed for cleaning.
  • Airflow channels for ventilation.
  • Absorbent terry toweling sweatband.
  • Available in different peak lengths for a variety of applications.
  • Adjustable elastic strap for a personal fit.
  • Optional logo customization areas.
  • Available in different peak lengths for a variety of applications.
  • Adjustable elastic strap for a personal fit.
  • Belt clip accessory available.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
3M™ First Base™ + Bump Cap > First Base™ + Bump Cap; Type: Standard Peak; Color: Navy Blue